Learn More
IL-38/IL-1F10 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2427
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417554
  • 486.22 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
IL-38/IL-1F10 Polyclonal antibody AF2427 specifically detects IL-38/IL-1F10 in Human samples. It is validated for Western Blotting
Spezifikation
Spezifikation
IL-38/IL-1F10
LYOPH
Western Blot 0.1 ug/mL
Q8WWZ1
FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human IL-38/IL-1F10 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) IL-1ra and rhIL-1F5 is observed and less than 1% cross-reactivity with rhIL-1 alpha , rhIL-1 beta , rhIL-18, rhIL-1F6, rhIL-1F7, rhIL-1F8, and rhIL-1 F9 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL1F10
E. coli-derived recombinant human IL-38/IL-1F10 Cys2-Trp152 Accession # Q8WWZ1
Antigen Affinity-purified
RUO
Primary
84639
Reconstitute at 0.2 mg/mL in sterile PBS.