missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human Kappa Light Chain Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen A18855
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Human Kappa Light Chain Polyclonal antibody specifically detects Human Kappa Light Chain in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Spezifikation
Human Kappa Light Chain | |
Polyclonal | |
Unconjugated | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
> 4.5 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
Human Kappa Light Chain | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |