missing translation for 'onlineSavingsMsg'
Learn More

Laminin alpha 1 N-Terminus Domain VI Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Marke:  RD Systems (Bio-Techne) AF4187

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13418604

  • 351.13 € / 100 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Laminin alpha 1 N-Terminus Domain VI Polyclonal antibody AF4187 Specifically detects Laminin alpha 1 N-Terminus Domain VI in Human samples. It is validated for Immunohistochemistry, Western Blotting
Spezifikation

Spezifikation

Laminin alpha 1
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
P25391
LAMA, LAMA1, LAMA1 laminin, alpha 1, S-LAM-alpha
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human Laminin alpha 1 N-Terminus Domain VI in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse LAMA4 is observed.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
LAMA1
E. coli-derived recombinant human Laminin alpha 1 N-Terminus Domain VI, Leu22-Met269, Accession # P25391
Immunogen affinity purified
RUO
Primary
284217
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only