Learn More
Methionine Aminopeptidase 1/METAP1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF3537
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418224
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 30-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Methionine Aminopeptidase 1/METAP1 Polyclonal antibody AF3537 Specifically detects Methionine Aminopeptidase 1/METAP1 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Spezifikation
Spezifikation
Methionine Aminopeptidase 1/METAP1
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
P53582
DKFZp781C0419, EC 3.4.11.18, KIAA0094MAP 1, MAP1, MAP1A, MetAP 1, MetAP1, MetAP1A, methionine aminopeptidase 1, methionyl aminopeptidase 1, Peptidase M 1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Methionine Aminopeptidase 1/METAP1 in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse METAP1 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
METAP1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Methionine Aminopeptidase 1/METAP1 His52-Phe386 Accession # P53582
Immunogen affinity purified
RUO
Primary
23173
Reconstitute at 0.2 mg/mL in sterile PBS.