Learn More
HAND2 Goat anti-Human, Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF3876
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418424
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 31-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
HAND2 Polyclonal antibody AF3876 Specifically detects HAND2 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Spezifikation
Spezifikation
HAND2
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
P61296
BHLHA26, bHLHa26MGC125303, dHAND, DHAND2, Ehand2, FLJ16260, heart and neural crest derivatives expressed 2, heart, autonomic nervous system and neural crestderivatives-expressed protein 2, Hed, MGC125304, Th2, Thing2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse HAND2 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human HAND1 is observed.
Human, Mouse
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
HAND2
E. coli-derived recombinant human HAND2 Met1-Gln217 Accession # P61296
Immunogen affinity purified
RUO
Primary
9464
Reconstitute at 0.2 mg/mL in sterile PBS.