Learn More
IL-17B Goat anti-Human, Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF1248
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13416844
  • 351.13 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
IL-17B Polyclonal antibody AF1248 Specifically detects IL-17B in Human, Mouse samples. It is validated for Western Blotting
Spezifikation
Spezifikation
IL-17B
LYOPH
Western Blot 0.1 ug/mL
Q9UHF5
Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL17B, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse IL-17B in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant human (rh) IL-17, rhIL-17C, rhIL-17E, and rhIL-17F is observed.
Human, Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL17B
E. coli-derived recombinant human IL-17B (R&D Systems, Catalog # 1248-IB), Gln21-Phe180, Accession # Q9UHF5
Immunogen affinity purified
RUO
Primary
27190
Reconstitute at 0.2 mg/mL in sterile PBS.