Learn More
OSCAR Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2004
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417284
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 22-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
OSCAR Polyclonal antibody AF2004 Specifically detects OSCAR in Human samples. It is validated for Western Blotting
Spezifikation
Spezifikation
OSCAR
LYOPH
Western Blot 0.1 ug/mL
AAM19095
hOSCAR, MGC33613, osteoclast associated receptor OSCAR-S1, osteoclast associated receptor OSCAR-S2, osteoclast associated, immunoglobulin-like receptor, osteoclast-associated immunoglobulin-like receptor, Osteoclast-associated receptor, PIgR-3, PIGR3, Poly-Ig receptor 3, Polymeric immunoglobulin receptor 3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human OSCAR in direct ELISAs and Western blots. In these formats, approximately 25% cross-reactivity with recombinant mouse OSCAR is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
OSCAR
Mouse myeloma cell line NS0-derived recombinant human OSCAR isoform 2, Asp19-Asn229 (Asn158Ser), Accession # AAM19095
Immunogen affinity purified
RUO
Primary
126014
Reconstitute at 0.2 mg/mL in sterile PBS.