Learn More
HPV Monoclonal Antibody (B73H), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA17054
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-7054 detects the human Pampilloma virus. MA1-7054 has been successfully used in ELISA and Western blot procedures. The MA1-7054 immunogen is purified recombit E7 protein of HPV types 16 and 18.
Papillomaviruses are a diverse group of DNA-based viruses that infect the skin and mucous membranes of humans and a variety of animals. 100 different HPV types have been identified to date. Various types of HPV may cause warts while others may cause a subclinical infection resulting in precancerous lesions. All HPVs are transmitted by skin-to-skin contact.Spezifikation
HPV | |
Monoclonal | |
0.1 mg/mL | |
Human papilloma virus | |
Purified recombit E7 protein. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
B73H | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |