Learn More
R-Spondin 2 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF3266
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418074
  • 351.13 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
R-Spondin 2 Polyclonal antibody AF3266 Specifically detects R-Spondin 2 in Human samples. It is validated for Western Blotting
Spezifikation
Spezifikation
R-Spondin 2
LYOPH
Western Blot 0.1 ug/mL
Q6UXX9
Cristin 2, CRISTIN2, hRspo2, MGC35555, MGC43342, Roof plate-specific spondin-2, RSPO2, RSpondin 2, R-spondin 2 homolog (Xenopus laevis), R-spondin-2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human R-Spondin 2 in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant human (rh) R-Spondin 1, rhR-Spondin 3, and recombinant mouse R-Spondin 4 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
RSPO2
E. coli-derived recombinant human R-Spondin 2 isoform 1 Cys21-Gly205 Accession # Q6UXX9
Immunogen affinity purified
RUO
Primary
340419
Reconstitute at 0.2 mg/mL in sterile PBS.