Learn More
Serpin C1/Antithrombin-III Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF1267
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13416854
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 04-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Serpin C1/Antithrombin-III Polyclonal antibody AF1267 Specifically detects Serpin C1/Antithrombin-III in Human samples. It is validated for Immunoprecipitation, Neutralization, Western Blotting
Spezifikation
Spezifikation
Serpin C1/Antithrombin-III
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 4.6 ug/mL
P01008
Antithrombin-III, AT3antithrombin-III, ATIII, ATIIIantithrombin III, MGC22579, serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1, serine-cysteine proteinase inhibitor clade C member 1, Serpin C1, serpin peptidase inhibitor, clade C (antithrombin), member 1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Serpin C1/Antithrombin-III in direct ELISAs and Western blots. In direct ELISAs, approximately 45% cross-reactivity with recombinant mouse (rm) Serpin C1 is observed and less than 2% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A4, rhSerpin A5, rhSerpin F2, rmSerpin D1 and rhPEDF is observed.
Human
Western Blot, Immunoprecipitation, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SERPINC1
Mouse myeloma cell line NS0-derived recombinant human Serpin C1/Antithrombin-III His33-Lys464 Accession # P01008
Antigen Affinity-purified
RUO
Primary
462
Reconstitute at 0.2 mg/mL in sterile PBS.