Learn More
Human Serum Albumin Monoclonal Antibody (1E1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA514698
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-14698 targets Human Serum Albumin in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MA5-14698 immunogen is human Serum Albumin. MA5-14698 detects Human Serum Albumin which has a predicted molecular weight of approximately 67 kDa. Product MA514698 is a smaller package size of MIH3001 (formerly sold as a Seradyn product).
Human Serum Albumin Monoclonal antibody specifically detects Human Serum Albumin in Human samples. It is validated for ELISA, Immunoprecipitation, Radioimmune Assays (RIA).Spezifikation
Human Serum Albumin | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
P02768 | |
ALB | |
Human Serum Albumin | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
1E1 | |
Unconjugated | |
ALB | |
Alb; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 | |
Mouse | |
Protein A | |
RUO | |
213 | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |