Learn More
Siglec-6/CD327 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2859
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417834
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 31-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Siglec-6/CD327 Polyclonal antibody AF2859 Specifically detects Siglec-6/CD327 in Human samples. It is validated for Western Blotting
Spezifikation
Spezifikation
Siglec-6/CD327
LYOPH
Western Blot 0.1 ug/mL
NP_942142
CD327, CD327 antigen, CD33 antigen-like 1, CD33L1, CD33L1OBBP1CD327, CD33L2, CD33LCDW327, CDw327, OB-BP1, Obesity-binding protein 1, sialic acid binding Ig-like lectin 6, sialic acid-binding Ig-like lectin 6, Siglec6, Siglec-6
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Siglec-6/CD327 in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human (rh) Siglec-3, rhSiglec-5 and rhSiglec-9 is observed, approximately 5% cross-reactivity with rhSiglec-10 is observed, and less than 1% cross-reactivity with rhSiglec-2, rhSiglec-7, and rhSiglec-11 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SIGLEC6
Chinese hamster ovary cell line CHO-derived recombinant human Siglec-6/CD327 Gln27-Val331 Accession # NP_942142
Immunogen affinity purified
RUO
Primary
946
Reconstitute at 0.2 mg/mL in sterile PBS.