Learn More
TRIM5 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF3684
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418314
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
TRIM5 Polyclonal antibody AF3684 Specifically detects TRIM5 in Human samples. It is validated for Western Blotting
Spezifikation
Spezifikation
TRIM5
LYOPH
Western Blot 0.1 ug/mL
Q9C035
EC 6.3.2, EC 6.3.2.-, RING finger protein 88, RNF88, RNF88tripartite motif protein TRIM, TRIM5alpha, tripartite motif containing 5, tripartite motif protein TRIM5, tripartite motif-containing 5, tripartite motif-containing protein 5
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human TRIM5 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse TRIM5 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TRIM5
E. coli-derived recombinant human TRIM5, Ala2-Gln248, Accession # Q9C035
Immunogen affinity purified
RUO
Primary
85363
Reconstitute at 0.2 mg/mL in sterile PBS.