Learn More
ULBP-1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF1380
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13416914
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
ULBP-1 Polyclonal antibody AF1380 Specifically detects ULBP-1 in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Spezifikation
Spezifikation
ULBP-1
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
Q9BZM6
ALCAN-beta, MULT-1, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL, NKG2DL1, RAET1I, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1, ULBP1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human ULBP-1 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) ULBP-2 and rhULBP-3 is observed.
Human
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ULBP1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ULBP-1, Gly26-Pro215, Accession # Q9BZM6
Antigen Affinity-purified
RUO
Primary
80329
Reconstitute at 0.2 mg/mL in sterile PBS.