Learn More
ULBP-3 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF1517
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13416984
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
ULBP-3 Polyclonal antibody AF1517 Specifically detects ULBP-3 in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Spezifikation
Spezifikation
ULBP-3
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
NP_078794
NKG2D ligand 3, RAET1N, UL16 binding protein 3, ULBP3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human ULBP-3 in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant human (rh) ULBP-2 is observed and less than 1% cross-reactivity with rhULBP-1 is observed.
Human
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ULBP3
Mouse myeloma cell line NS0-derived recombinant human ULBP-3, Gly27-Pro216, Accession # NP_078794
Immunogen affinity purified
RUO
Primary
79465
Reconstitute at 0.2 mg/mL in sterile PBS.