Learn More
Vasorin/SLIT-like 2 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Marke:  RD Systems (Bio-Techne) AF2140
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417374
  • 479.72 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Vasorin/SLIT-like 2 Polyclonal antibody AF2140 Specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for Western Blotting
Spezifikation
Spezifikation
Vasorin/SLIT-like 2
LYOPH
Western Blot 0.1 ug/mL
Affinity Purified
VASN
Chinese hamster ovary cell line CHO-derived recombinant human Vasorin extracellular domain
Immunogen affinity purified
RUO
Primary
114990
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Atia, Protein slit-like 2, SLIT like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), VASN, vasorin
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Vasorin in direct ELISAs and Western blots.