Learn More
Huntingtin Monoclonal Antibody (HDB4E10), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516703
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Increased cytoplasmic staining, relative to nuclear, has been reported using formaldehyde as a fixative compared with acetone/methanol. A suggested positive control for immunohistochemical applications is brain.
Huntingtin Monoclonal antibody specifically detects Huntingtin in Human, Mouse, Rabbit samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Spezifikation
Huntingtin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P42858, P42859 | |
Htt | |
Recombinant protein corresponding to amino acids 1844 - 2131 of huntingtin. | |
100 μg | |
Primary | |
Human, Mouse, Rabbit | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
HDB4E10 | |
Unconjugated | |
Htt | |
AI256365; C430023I11Rik; Hd; HD protein; HD protein homolog; Hdh; HTT; Huntingtin; huntingtin (Huntington disease); Huntingtin, myristoylated N-terminal fragment; Huntington disease gene homolog; huntington disease protein; huntington disease protein homolog; IT15; solute carrier family 6 (neurotransmitter transporter, serotonin), member 4 | |
Mouse | |
purified | |
RUO | |
100351801, 15194, 3064 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |