missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HUS1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543917
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: ELFIHVSGTV ARLAKVCVLR VRPDSLCFGP AGSGGLHEAR LWCEVRQGAF Sequence homology: Human: 100%
HUS1B Polyclonal antibody specifically detects HUS1B in Human samples. It is validated for Western Blot.Spezifikation
HUS1B | |
Polyclonal | |
Unconjugated | |
Hus1b | |
Checkpoint protein HUS1B; hHUS1B; HUS1 checkpoint clamp component B; HUS1 checkpoint homolog b; HUS1 checkpoint homolog b (S. pombe); Hus1 homolog b (S. pombe); Hus1b; MGC126746; MGC126748; mHUS1B; RP11-532F6.1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
135458 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NHY5 | |
Hus1b | |
Synthetic peptide directed towards the N-terminal of human HUS1B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |