Learn More
HK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528217
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (94%), Rat (92%), Zebrafish (84%), Xenopus laevis (86%), Pig (93%), Chicken (88%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HK1 Polyclonal antibody specifically detects HK1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HK1 | |
Polyclonal | |
Unconjugated | |
HK1 | |
BB404130; brain form hexokinase; dea; downeast anemia; glycolytic enzyme; HEXOKIN; hexokinase; hexokinase 1; hexokinase I; hexokinase IR; hexokinase type I; Hexokinase, tumor isozyme; hexokinase-1; Hexokinase-A; HK; HK I; HK1; Hk-1; Hk1-s; HK1-ta; HK1-tb; HK1-tc; HKD; HKI; HMSNR; HXK1; mHk1-s | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3098 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P19367 | |
HK1 | |
Recombinant fragment corresponding to a region within amino acids 529 and 852 of Human HXK I | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |