missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYOU1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA514203
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
HYOU1 Polyclonal antibody specifically detects HYOU1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HYOU1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9Y4L1 | |
HYOU1 | |
KLH conjugated synthetic peptide between 274-303 amino acids from the central region of human HYOU1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
HYOU1 | |
140 kDa Ca(2+)-binding protein; 150 kDa oxygen-regulated protein; 150-kDa oxygen regulated protein; 170 kDa glucose regulated protein GRP170; 170 kDa glucose-regulated protein; AI415631; Cab140; calcium binding protein 140; calcium binding protein, 140 kDa; CBP-140; DKFZp686N08236; FLJ94899; FLJ97572; Grp 170; Grp170; GRP-170; HSP 12A; HSP12A; HYOU1; hypoxia up-regulated 1; hypoxia up-regulated protein 1; Orp 150; Orp150; ORP-150; oxygen regulated protein (150kD) | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
10525 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |