missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRICH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528804
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: hypothetical protein LOC51233 tansfected 293T cell. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DRICH1 Polyclonal antibody specifically detects DRICH1 in Human samples. It is validated for Western Blot.Spezifikation
DRICH1 | |
Polyclonal | |
Unconjugated | |
DRICH1 | |
aspartate rich 1; aspartate-rich 1; aspartate-rich protein 1; C22orf43; DRICH1; KB-208E9.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51233 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q6PGQ1 | |
DRICH1 | |
Synthetic peptide corresponding to a region within amino acids 166 and 229 of Human hypothetical protein LOC51233 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |