missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGHA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA514361
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
IGHA1 Polyclonal antibody specifically detects IGHA1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
IGHA1 | |
Polyclonal | |
Unconjugated | |
IGHA1 | |
IgA1; IGHA1; IGHA2; immunoglobulin heavy constant alpha 1; immunoglobulin heavy constant alpha 2 (A2m marker) | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
3493 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2.0 mg/mL | |
PBS with 0.09% sodium azide | |
P01876 | |
IGHA1 | |
KLH conjugated synthetic peptide between 257-286 amino acids from the C-terminal region of human IGHA1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |