missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKAROS Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519214
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine and canine based on sequence homology.
IKAROS Polyclonal antibody specifically detects IKAROS in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
IKAROS | |
Polyclonal | |
Unconjugated | |
IKZF1 | |
5832432G11Rik; CLL-associated antigen KW-6; CVID13; DNA-binding protein Ikaros; hIk-1; hlk-1; Hs.54452; IK1; IKAROS; IKAROS family zinc finger 1; IKAROS family zinc finger 1 (Ikaros); ikaros family zinc finger protein 1; IKZF1; KAROS family zinc finger 1 (Ikaros); LYF1; LyF-1; Lymphoid transcription factor LyF-1; mKIAA4227; PPP1R92; PRO0758; protein phosphatase 1, regulatory subunit 92; RGD1562979; Zfpn1a1; zinc finger containing transcription factor; zinc finger protein, subfamily 1A, 1 (Ikaros); Znfn1a1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10320 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q13422 | |
IKZF1 | |
Synthetic peptide sequence (DTPYDSSASYEKEN) corresponding to the internal amino acids of IKZF1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |