missing translation for 'onlineSavingsMsg'
Learn More

IKK beta Monoclonal Antibody (10A2C5B3), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA515320

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11395293

  • 463.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA5-15320 targets IKBKB in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15320 immunogen is purified recombinant fragment of IKBKB expressed in E. Coli.

IKK beta Monoclonal antibody specifically detects IKK beta in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.
Spezifikation

Spezifikation

IKK beta
Monoclonal
Conc. Not Determined
ascites with 0.03% sodium azide
O14920
Ikbkb
Purified recombinant fragment of IKBKB expressed in E. Coli.
RUO
3551
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry, Western Blot
10A2C5B3
Unconjugated
Ikbkb
AI132552; AIM-1; FLJ40509; I-kappa-B kinase 2; I-kappa-B-kinase beta; Ikbkb; IKK[b]; IKK2; IKK-2; IKKB; IKK-B; IKKbeta; IKK-beta; IMD15; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta; inhibitor of kappaB kinase beta; inhibitor of nuclear factor kappa B kinase beta subunit; inhibitor of nuclear factor kappa-B kinase subunit beta; MGC131801; NFKBIKB; nuclear factor NF-kappa-B inhibitor kinase beta
Mouse
100 μL
Primary
Human
Antibody
IgG1
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote