Learn More
IL-1 alpha Monoclonal Antibody (40508), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA523736
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In ELISAs, this antibody shows less than 1% cross-reactivity with recombinant rat (rr) IL-1 alpha /IL-1F1, rpIL-1 alpha /IL-1F1, or rhIL-1 alpha /IL-1F1. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL-1 alpha Monoclonal antibody specifically detects IL-1 alpha in Mouse samples. It is validated for ELISA, Neutralization.Spezifikation
IL-1 alpha | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P01582 | |
Il1a | |
E. coli-derived recombinant mouse IL-1 alpha/IL-1F1 Ser115-Ser270 | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
ELISA, Neutralization | |
40508 | |
Unconjugated | |
Il1a | |
CTLA8; hematopoietin-1; IL 1 a; IL 1a; IL1; IL-1 alpha; IL17; IL1A; Il-1a; IL-1alpha; IL1-ALPHA; IL1F1; IL1a; ILN; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin 17; interleukin 1a; interleukin 1-alpha; interleukin 1-alpha (AA 1 - 152); Interleukin1 alpha; interleukin-1 alpha; interleukin-1 alpha precursor; Interleukin-17A; preinterleukin 1 alpha; pre-interleukin-1 alpha; pro-interleukin-1-alpha; RP23-160G19.8 | |
Rat | |
Protein A/G | |
RUO | |
16175 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |