Learn More
IL-1 alpha Recombinant Polyclonal Antibody (1HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Marke: Invitrogen 710043
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
IL-1 alpha Recombinant Polyclonal antibody specifically detects IL-1 alpha in Human samples. It is validated for Western Blot.Spezifikation
IL-1 alpha | |
Recombinant Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P01583 | |
IL1A | |
Protein corresponding to amino acids 113-271 of human IL-1 alpha | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
1HCLC | |
Unconjugated | |
IL1A | |
CTLA8; hematopoietin-1; IL 1 a; IL 1a; IL1; IL-1 alpha; IL17; IL1A; Il-1a; IL-1alpha; IL1-ALPHA; IL1F1; IL1a; ILN; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin 17; interleukin 1a; interleukin 1-alpha; interleukin 1-alpha (AA 1 - 152); Interleukin1 alpha; interleukin-1 alpha; interleukin-1 alpha precursor; Interleukin-17A; preinterleukin 1 alpha; pre-interleukin-1 alpha; pro-interleukin-1-alpha; RP23-160G19.8 | |
Rabbit | |
Protein A | |
RUO | |
3552 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |