Learn More
IL-1 beta Monoclonal Antibody (1400.24.17), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MM425BB
Weitere Details : Netto-Gewicht : 0.80000kg
Descripción
MM425BB targets IL-1 beta in ELISA, and WB applications and shows reactivity with mouse samples. The MM425BB immunogen is recombinant mouse IL-1 beta. MM425BB detects IL-1 beta which has a predicted molecular weight of approximately 17 kDa.
IL-1 beta Monoclonal antibody specifically detects IL-1 beta in Mouse samples. It is validated for ELISA, Western Blot.Especificaciones
IL-1 beta | |
Monoclonal | |
0.5 mg/mL | |
PBS with 4% BSA and proprietary preservative | |
P10749 | |
IL1B | |
Recombinant mouse IL-1 beta | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
1400.24.17 | |
Biotin | |
IL1B | |
catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta | |
Mouse | |
Protein A | |
RUO | |
16176 | |
-20°C | |
Liquid |