missing translation for 'onlineSavingsMsg'
Learn More

IL-1 beta Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Marke:  Invitrogen PA546956

Weitere Details : Netto-Gewicht : 0.00010kg

Artikelnummer. 15507452

  • 516.00 € / 100 Mikrogramm
Verfügbar ab: 24-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

In sandwich ELISAs, less than 1% cross-reactivity with recombinant mouse IL-1 beta is observed and less than 0.5% cross-reactivity with recombinant human IL-1 beta and recombinant porcine IL-1 beta is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

IL-1 beta Polyclonal antibody specifically detects IL-1 beta in Rat samples. It is validated for ELISA, Immunocytochemistry, Neutralization, Western Blot.
Spezifikation

Spezifikation

IL-1 beta
Polyclonal
Unconjugated
IL1B
catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta
Goat
Antigen affinity chromatography
RUO
24494
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Immunocytochemistry, Neutralization, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
Q63264
IL1B
E. coli-derived recombinant rat IL-1 beta/IL-1F2 Val117-Ser268
100 μg
Primary
Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote