Learn More
IL1R1 Monoclonal Antibody (IL1 31-113.1.1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA110859
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-10859 detects IL-1 Receptor from human samples. MA1-10859 has been successfully used in ELISA and immunohistochemistry applications. The MA1-10859 immunogen is recombinant extracellular domain of the type I human IL-1 receptor.
IL1R1 Monoclonal antibody specifically detects IL1R1 in Human samples. It is validated for ELISA, Immunohistochemistry.Spezifikation
IL1R1 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P14778 | |
Il1r1 | |
Recombinant extracellular domain of the type I human IL-1 receptor. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Immunohistochemistry | |
IL1 31-113.1.1 | |
Unconjugated | |
Il1r1 | |
antigen CD121a; CD121 alpha; CD121 antigen-like family member A; CD121a; CD121b; D2S1473; il1 receptor; il-1 receptor; IL-1 receptor alpha chain; IL1R; il-1r; IL1R1; IL-1R1; IL-1R-1; Il1r-1; IL1RA; IL-1R-alpha; IL1RT1; IL-1RT1; IL-1RT-1; IL-iR; interleukin 1 beta receptor type 1 short transmembrane form; interleukin 1 beta receptor type 1 soluble form; interleukin 1 bete receptor type 1 long transmembrane form; interleukin 1 receptor alpha, type I; interleukin 1 receptor type 1; interleukin 1 receptor, type I; interleukin-1 receptor; interleukin-1 receptor alpha; Interleukin-1 receptor type 1; Interleukin-1 receptor type 1, membrane form; Interleukin-1 receptor type 1, soluble form; Interleukin-1 receptor type I; mIL-1R1; p80; sIL-1R1 | |
Mouse | |
Protein G | |
RUO | |
3554 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |