Learn More
IL13RA2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA546976
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse IL-13 R alpha 2 is observed and less than 1% cross-reactivity with recombinant human IL-13 R alpha 1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL13RA2 Polyclonal antibody specifically detects IL13RA2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Neutralization, Western Blot.Spezifikation
IL13RA2 | |
Polyclonal | |
Unconjugated | |
IL13RA2 | |
cancer/testis antigen 19; CD213a2; CT19; IL-13 receptor subunit alpha-2; IL13BP; IL13R; IL-13R; IL-13R subunit alpha-2; IL13RA2; IL-13RA2; IL-13R-alpha-2; interleukin 13 binding protein; interleukin 13 receptor alpha 2; interleukin 13 receptor alpha 2 chain; interleukin 13 receptor alpha chain 2; interleukin 13 receptor subunit alpha 2; interleukin 13 receptor, alpha 2; interleukin-13 receptor subunit alpha-2; interleukin-13 receptor subunit alpha-2-like; interleukin-13-binding protein | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3598 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q14627 | |
IL13RA2 | |
Mouse myeloma cell line NS0-derived recombinant human IL-13 R alpha 2 Cys22-Leu342 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |