missing translation for 'onlineSavingsMsg'
Learn More

IL15RA Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Marke:  Invitrogen PA547749

292.68 EUR gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Weitere Details : Netto-Gewicht : 0.00010kg

Artikelnummer. 15538242

  • 439.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) IL-15 R alpha is observed and less than 1% cross-reactivity with rhIL-2 R alpha, recombinant mouse (rm) IL-2 R beta, and rmIL-2 R gamma is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

IL15RA Polyclonal antibody specifically detects IL15RA in Mouse samples. It is validated for Flow Cytometry, Neutralization, Western Blot.
Spezifikation

Spezifikation

IL15RA
Polyclonal
Unconjugated
IL15RA
AA690181; CD215; IL 15 receptor; IL 15 receptor subunit a; IL 15R; IL 15R alpha; IL 15R a; IL15 receptor; IL15 receptor subunit alpha; IL-15 receptor subunit alpha; IL15 receptor subunit a; IL15R; IL15R alpha; IL15R a; Il15ra; IL-15RA; IL-15R-alpha; Interleukin 15 receptor; interleukin 15 receptor alpha isoform EM2; interleukin 15 receptor alpha isoform IC2; interleukin 15 receptor alpha isoform IC3; interleukin 15 receptor alpha isoform IC4; interleukin 15 receptor alpha isoform IC5; interleukin 15 receptor alpha isoform IC6; interleukin 15 receptor alpha isoform IC7; interleukin 15 receptor alpha isoform IC8; interleukin 15 receptor subunit alpha; Interleukin 15 receptor subunit a; interleukin 15 receptor, alpha; interleukin 15 receptor, alpha chain; interleukin-15 receptor subunit alpha; sIL 15R; sIL-15 receptor subunit alpha; sIL15R; sIL-15RA; sIL-15R-alpha; soluble IL 15R; soluble IL15R; Soluble interleukin-15 receptor subunit alpha
Goat
Antigen affinity chromatography
RUO
16169
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Flow Cytometry, Neutralization, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
Q60819
IL15RA
Mouse myeloma cell line NS0-derived recombinant mouse IL-15 R alpha Gly33-Lys205
100 μg
Primary
Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote