missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL17RA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534571
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is A-20 cell lysate. PA5-34571 can be used with blocking peptide PEP-1614.
IL17RA Polyclonal antibody specifically detects IL17RA in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
IL17RA | |
Polyclonal | |
Unconjugated | |
Il17ra | |
AW538159; CANDF5; CD127; CD217; CDw217; hIL-17R; IL-17 receptor A; IL17R; Il17ra; IL-17RA; interleukin 17 receptor A; interleukin-17 receptor A; MGC10262; VDw217 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16172, 23765, 312679 | |
4° C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q60943, Q96F46 | |
Il17ra | |
An 18 amino acid peptide near the amino terminus of human IL-17RA | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |