missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL18R1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA124812
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IL18R1 Polyclonal antibody specifically detects IL18R1 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
IL18R1 | |
Polyclonal | |
Unconjugated | |
Il18r1 | |
CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha | |
Goat | |
Antigen affinity chromatography | |
RUO | |
8809 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
0.1 mg/mL | |
PBS with no preservative | |
Q13478 | |
Il18r1 | |
Recombinant, full length human IL18R1 protein expressed in mouse NSO cells. | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |