Learn More
IL-2 Rat anti-Mouse, PE, Clone: JES6-5H4, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 561061
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The JES6-5H4 monoclonal antibody specifically binds to mouse interleukin-2 (IL-2). IL-2 is a multifunctional cytokine that plays pivotal roles in immunity and tolerance. It is produced by activated T cells. IL-2 effects the activation, growth, proliferation and/or differentiation of various cell types including T and B lymphocytes and their precursors, LAK cells, NK cells, and monocytes/macrophages. IL-2 mediates its biological activities by binding to IL-2 receptor compleses. The intermediate affinity IL-2R is comprised of IL-2R β (CD122) and common gamma chain ( γc; CD132) subunits whereas the high-affinity IL-2R is comprised of IL-2R α (CD25), IL-2R β and γc subunits. The JES6-5H4 monoclonal antibody binds to IL-2 and neutralizes its biological activity.
Spezifikation
IL-2 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse IL-2 Recombinant Protein | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
JES6-5H4 | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2b |
For Research Use Only.