missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen ARC0022
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Spezifikation
IL-2 | |
Polyclonal | |
Il2 | |
T-cell growth factor, TCGF, interleukin-2 | |
Rabbit | |
Protein A/G | |
RUO | |
116562 | |
-20°C | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
P17108 | |
Il2 | |
Purified recombit rat IL-2. | |
500 μg | |
Primary | |
Rat | |
Antibody | |
IgG |