missing translation for 'onlineSavingsMsg'
Learn More

IL-21 Recombinant Polyclonal Antibody (18HCLC), Invitrogen™

Rabbit Recombinant Polyclonal Antibody

Marke:  Invitrogen 710141

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15307214

  • 470.00 € / 100 Mikrogramm
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.


Beschreibung

Beschreibung

Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.

IL-21 Recombinant Polyclonal antibody specifically detects IL-21 in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

IL-21
Polyclonal
0.5 mg/mL
PBS with 0.09% sodium azide
CVID11; CX2; il 21; IL21; IL-21; ILN; Interleukin; interleukin 21; Interleukin21; interleukin-21; interleukin-21 isoform; porcine interleukin-21; Za11
Recombinant protein corresponding to amino acids 37-162 of human interleukin-21
100 μg
Primary
Human
Liquid
Western Blot
18HCLC
Unconjugated
Q9HBE4
Rabbit
Protein A
RUO
59067
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote