missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-22 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA121356
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Human spleen and tonsil.
IL-22 Polyclonal antibody specifically detects IL-22 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Spezifikation
IL-22 | |
Polyclonal | |
Unconjugated | |
IL22 | |
Cytokine Zcyto18; EGK_03903; il 22; IL-10-related T-cell-derived inducible factor; IL-10-related T-cell-derived-inducible factor; IL-21; IL22; IL-22; Il22a; IL-22a; IL-D110; ILN; ILTIF; IL-TIF; ILTIF alpha; IL-TIF alpha; ILTIFa; Interleukin; interleukin 10-related T cell-derived inducible factor; interleukin 22; Interleukin22; interleukin-22; Interleukin-22a; RGD1561292; TIFa; TIFIL-23; UNQ3099/PRO10096; ZCYTO18 | |
Goat | |
Affinity chromatography | |
RUO | |
50616, 50929 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
0.01M potassium phosphate with 0.14M NaCl, 1mg/mL BSA and 0.1% sodium azide | |
Q9GZX6, Q9JJY9 | |
IL22 | |
Synthetic peptide of Human IL22 C terminal amino acids 148-177 [LKDTVKKLGESGEIKAIGELDLLFMSLRNA] | |
50 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |