Learn More
IL-23 p19 Recombinant Polyclonal Antibody (26HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Marke: Invitrogen 710259
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with non-human primate, mouse and rat based on sequence homology. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
IL-23 p19 Recombinant Polyclonal antibody specifically detects IL-23 p19 in Human samples. It is validated for Western Blot.Spezifikation
IL-23 p19 | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
il 23; IL 23 A; IL12B; Il-12b; Il12p40; Il-12p40; il23; IL-23; IL-23 subunit alpha; IL23A; IL-23A; IL-23-A; IL23P19; IL-23p19; ILN; Interleukin; interleukin 12B; interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; MGC79388; P19; p40; RP23-388G23.1; SGRF; UNQ2498/PRO5798 | |
Peptide corresponding to amino acids 55–67 of human IL-23 | |
100 μg | |
Primary | |
Human | |
Liquid |
Western Blot | |
26HCLC | |
Unconjugated | |
Q9NPF7 | |
Rabbit | |
Protein A | |
RUO | |
51561 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |