missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL24 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA141066
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Suggested positive control: Jurkat, antigen standard for IL24 (transient overexpression lysate), Jurkat whole cell lysate.
IL24 Polyclonal antibody specifically detects IL24 in Human, Non-human Primate samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
IL24 | |
Polyclonal | |
Unconjugated | |
IL24 | |
C49A; Cytokine-like protein Mob-5; FISP; IL 24; IL10B; Il24; IL-24; IL-4-induced secreted protein; interleukin 24; interleukin-24; Mda7; MDA-7; melanocyte-associated Mda-7; melanoma differentiation-associated gene 7 protein; Mob5; ST16; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein; Th2-specific cytokine FISP | |
Rabbit | |
Protein G | |
RUO | |
11009 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.2% gelatin and 0.05% sodium azide | |
Q13007 | |
IL24 | |
udSynthetic peptide corresponding to aa 123-136 of human IL-24. | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |