missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFNLR1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA121360
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive control is mouse spleen.
IFNLR1 Polyclonal antibody specifically detects IFNLR1 in Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Spezifikation
IFNLR1 | |
Polyclonal | |
Unconjugated | |
Ifnlr1 | |
class II cytokine receptor 12; class II cytokine receptor CRF2/12; CRF2/12; CRF2-12; Cytokine receptor class-II member 12; cytokine receptor family 2 member 12; IFN-lambda R1; IFN-lambda receptor 1; IFN-lambda-R1; IFNLR; IFNLR1; IL-28 receptor subunit alpha; IL-28R1; Il28ra; IL-28RA; IL-28R-alpha; Interferon lambda receptor 1; interferon lambda, receptor 1; interferon, lambda receptor 1; interferon-lambda receptor 1; interleukin 28 alpha receptor; interleukin 28 receptor A; interleukin 28 receptor alpha; interleukin 28 receptor, alpha (interferon, lambda receptor); interleukin or cytokine receptor 2; Interleukin-28; interleukin-28 receptor subunit alpha; LICR2; Likely interleukin or cytokine receptor 2; RGD1562689 | |
Goat | |
Affinity chromatography | |
RUO | |
242700, 362628 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
0.01M potassium phosphate with 0.14M NaCl, 1mg/mL BSA and 0.1% sodium azide | |
Q8CGK5 | |
Ifnlr1 | |
Synthetic peptide of Mouse IL28 Receptor alpha N terminal a.a. 44-72 [LPGLGSPPNVTYFVTYQSYIKTGWRPVEH] | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |