Learn More
IL-29 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547165
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human (rh) IL-28A and rhIL-28B is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL-29 Polyclonal antibody specifically detects IL-29 in Human samples. It is validated for Neutralization, Western Blot.Spezifikation
IL-29 | |
Polyclonal | |
Unconjugated | |
IFNL1 | |
cytokine Zcyto21; Cytokine Zcyto22; FNL1; IFN lambda1; IFNL1; IFNL3; IFN-lambda-1; IFN-lambda-3; IFN-lambda-4; IL28B; IL-28B; IL28C; IL-28C; IL29; IL-29; ILN; interferon lambda 1; interferon lambda 3; Interferon lambda1; interferon lambda-1; Interferon lambda-3; interferon, lambda 1; interferon, lambda 3; interferon, lambda 4; interferon-lambda-1; Interleukin; interleukin 29 (interferon, lambda 1); interleukin-28B; Interleukin-28C; interleukin-29; ZCYTO21; ZCYTO22 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
282618 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8IU54 | |
IFNL1 | |
Mouse myeloma cell line NS0-derived recombinant human IL-29/IFN-lambda 1 Gly20-Thr200 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |