Learn More
IL-3 Monoclonal Antibody (4806), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA523808
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-3 is observed. This antibody does not neutralize the biological activity of recombinant mouse IL-3. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL-3 Monoclonal antibody specifically detects IL-3 in Human samples. It is validated for Immunocytochemistry, Neutralization, Western Blot.Spezifikation
IL-3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P08700 | |
IL3 | |
E. coli-derived recombinant human IL-3 Ala20-Phe152 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Neutralization, Western Blot | |
4806 | |
Unconjugated | |
IL3 | |
BP/BPA; BPA; Colony stimulating factor multiple; colony-stimulating factor, multiple; Csfmu; Eo-CSF; HCGF; hematopoietic growth factor; Il3; Il-3; ILN; Interleukin; interleukin 3; Interleukin 3 (colony stimulating factor; Interleukin3; interleukin-3; Mast cell growth factor; mast-cell growth factor; MCGF; MGC79398; MGC79399; MGF; Multi CSF; MULTI-CSF; Multilineage colony stimulating factor; multilineage-colony-stimulating factor; multiple); multipotential colony-stimulating factor; P cell stimulating factor; P-cell stimulating factor; P-cell-stimulating factor; PCSA; PSF | |
Mouse | |
Protein A/G | |
RUO | |
3562 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |