Learn More
IL-36Ra/IL-1F5 Mouse anti-Human, Clone: 190524, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB1275
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418515
  • 377.53 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 25-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
IL-36Ra/IL-1F5 Monoclonal antibody MAB1275 Specifically detects IL-36Ra/IL-1F5 in Human samples. It is validated for Western Blotting
Spezifikation
Spezifikation
IL-36Ra/IL-1F5
190524
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL36RN
E. coli-derived recombinant human IL-36Ra/IL-1F5 Met1-Asp155 Accession # Q9UBH0
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
26525
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9UBH0
FIL1 delta, IL1F5, IL-1HY1, IL-1L1, IL-1RP3, IL36Ra, IL-36RA, IL36RN, Interleukin 36 Receptor Antagonist, Interleukin-36 Receptor Antagonist
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human IL-36Ra/IL-1F5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) IL-1 alpha , rhIL-1 beta , rhIL-1ra, rhIL-18, rhIL-1F6, or rhIL-1F8.