missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD123 Mouse anti-Human, PE, Clone: 6H6, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen A16399
Weitere Details : Netto-Gewicht : 0.00280kg
Spezifikation
CD123 | |
Monoclonal | |
PE | |
RP11-261P4.2, CD123, IL3R, IL3RAY, IL3RX, IL3RY, hIL-3Ra | |
Mouse | |
RUO | |
3563 | |
Store at 2 - 8 °C. | |
IgG1 κ |
Flow Cytometry | |
6H6 | |
PBS, pH 7.2, with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide | |
IL3RA | |
25 Tests | |
Primary | |
Human | |
Liquid |