Learn More
IL-4 Mouse anti-Porcine, Clone: 99605, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB6543
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411416
  • 278.53 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
IL-4 Monoclonal antibody MAB6543 Specifically detects IL-4 in Porcine samples. It is validated for Flow Cytometry (Intracellular Staining), Western Blotting
Spezifikation
Spezifikation
IL-4
99605
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL4
E. coli-derived recombinant porcine IL-4 His25-Cys133 Accession # Q04745
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3565
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, CyTOF
LYOPH
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q04745
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects porcine IL-4 in direct ELISAs and Western blots. Does not cross-react with recombinant human IL-13 or with recombinant IL-4 from human, canine, bovine, mouse, rat, cotton rat, or feline systems.
Porcine