missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4 Monoclonal Antibody (A155B16F2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen ASC0944
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
This product is recommended as a capture antibody in Sandwich ELISA applications. This clone can also be referred to as clone number A155B 16F2 1A9.
IL-4 Monoclonal antibody specifically detects IL-4 in Porcine samples. It is validated for ELISA.Spezifikation
IL-4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.5mg/mL sodium azide; pH 7.5 | |
Q04745 | |
Il4 | |
Recombinant swine IL-4. | |
500 μg | |
Primary | |
Porcine | |
Antibody | |
IgG2b |
ELISA | |
A155B16F2 | |
Unconjugated | |
Il4 | |
B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra | |
Mouse | |
Protein A | |
RUO | |
397225 | |
4° C | |
Liquid |