Learn More
IL-6 Monoclonal Antibody (20F3), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MM600C
Weitere Details : Netto-Gewicht : 0.80000kg
Beschreibung
MM600C targets IL-6 in ELISA, IHC(P), IP, Neu, and WB applications and shows reactivity with mouse samples. The MM600C immunogen is recombinant mouse IL-6. MM600C detects IL-6 which has a predicted molecular weight of approximately 22 kDa. This antibody is produced by injecting Rat IgG secreting hybridoma cells into the peritoneum of mice. The resulting ascites is collected from the mice and the antibody is purified. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
IL-6 Monoclonal antibody specifically detects IL-6 in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Western Blot.Spezifikation
IL-6 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative | |
P08505 | |
Il6 | |
Recombinant mouse IL-6 | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Western Blot | |
20F3 | |
Unconjugated | |
Il6 | |
B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 | |
Rat | |
Protein G | |
RUO | |
16193 | |
-20°C | |
Liquid |