Learn More
IL-6 Receptor Monoclonal Antibody (255820), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA523951
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs and Western blots, no cross-reactivity with recombinant human IL-6 R is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL-6 Receptor Monoclonal antibody specifically detects IL-6 Receptor in Mouse samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
IL-6 Receptor | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P22272 | |
Il6ra | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-6 R alpha Leu20-Glu357 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
255820 | |
Unconjugated | |
Il6ra | |
CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; IL6R; IL-6R; IL-6R 1; IL-6R subunit alpha; IL6R1; IL-6R-1; Il6ra; IL-6RA; IL-6R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991 | |
Rat | |
Protein A/G | |
RUO | |
16194 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |