Learn More
IL-7 Monoclonal Antibody (7417), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA523700
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In ELISAs, does not cross-react with bovine FGF acidic, recombinant human (rh) G-CSF, rhGM-CSF, rhIL-1 alpha, rhIL-1 beta, rhIL-2, rhIL-3, rhIL-4, rhIL-6, recombinant mouse IL-7, rhIL-8, porcine PDGF, rhTNF-alpha, rhTNF-beta, human TGF-beta 1, or porcine TGF-beta 1. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL-7 Monoclonal antibody specifically detects IL-7 in Human samples. It is validated for ELISA, Neutralization, Western Blot.Spezifikation
IL-7 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P13232 | |
Il7 | |
E. coli-derived recombinant human IL-7 | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Neutralization, Western Blot | |
7417 | |
Unconjugated | |
Il7 | |
A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7 | |
Mouse | |
Protein A/G | |
RUO | |
3574 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |