Learn More
IL-8 (CXCL8) Monoclonal Antibody (8M6), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA182421
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody has also been reported to neutralize the bioactivity of ovine IL-8, however prior removal of sodium azide may be necessary for functional assays. Membrane permeabilization is required for flow cytometry applications. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Ovine Interleukin-8 antibody, clone 8M6 recognizes ovine interleukin-8 (IL-8), also known as C-X-C motif chemokine 8.
IL-8 (CXCL8) Monoclonal antibody specifically detects IL-8 (CXCL8) in Bovine, Canine, Ferret, Mink, Mustelid, Ovine, Porcine, Rabbit samples. It is validated for ELISA, Flow Cytometry, Functional Assay, Western Blot.Spezifikation
IL-8 (CXCL8) | |
Monoclonal | |
1 mg/mL | |
PBS with <0.1% sodium azide | |
P19874, P26894, P36925, P41324, P79255 | |
CXCL8 | |
Recombinant ovine IL-8 | |
250 μg | |
Primary | |
Bovine, Canine, Ferret, Mink, Mustelid, Ovine, Porcine, Rabbit | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Functional Assay, Western Blot | |
8M6 | |
Unconjugated | |
CXCL8 | |
(Ala-IL-8)77; (Ser-IL-8)72; Alveolar macrophage chemotactic factor I; alveolar macrophage-derived chemotactic factor-I; AMCF-I; ANX7; beta endothelial cell-derived neutrophil activating peptide; beta-thromboglobulin-like protein; chemokine (C-X-C motif) ligand 8; C-X-C motif chemokine 8; C-X-C motif chemokine ligand 8; CXCL8; cytokine 8; emoctakin; GCP/IL-8 protein I; GCP/IL-8 protein II; GCP/IL-8 protein III; GCP/IL-8 protein IV; GCP/IL-8 protein V; GCP/IL-8 protein VI; GCP1; GCP-1; Granulocyte chemotactic protein 1; IL8; IL-8; IL-8(1-77); IL-8(5-77); IL-8(6-77); IL-8(7-77); IL-8(8-77); IL-8(9-77); IL8/NAP1 form I; IL8/NAP1 form II; IL8/NAP1 form III; IL8/NAP1 form IV; IL8/NAP1 form V; IL8/NAP1 form VI; ILN; Interleukin; interleukin 8; Interleukin8; interleukin-8; interleuklin-8 precursor; LECT; LUCT; lung giant cell carcinoma-derived chemotactic protein; lymphocyte derived neutrophil activating peptide; Lymphocyte-derived neutrophil-activating factor; LYNAP; MDNCF; MDNCF-a; MDNCF-b; | |
Mouse | |
purified | |
RUO | |
100009129, 280828, 396880, 403850, 443418 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |